,,Multi-Element Solution 5 - 12 components: 10mg/l each of B Ge Mo Nb P Re S Si Ta Ti W Zr in HNO3 2% ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 36 months. (12 months after opening),, ,,Ambient Temp.,, 1 * 100 mL
Catalog # CMPLN9300235.L1
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together