Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Lutein Pharmaceutical Secondary Standard Certified Reference Material 1 * 3 items
Catalog # PHR1699-3X100M
Supplier:
Merck
Expand image
Lutein Pharmaceutical Secondary Standard Certified Reference Material 1 * 3 items
Catalog # PHR1699-3X100M
Supplier:
Merck
Frequently Bought Together
Nitrate standard solution, 1000 mg/L NO₃<sup>-</sup> in Water (from NaNO₃) Certipur®, Supelco®
Supplier: Merck
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Potassium hydrogen phthalate, Certipur® Reag. USP, Reag. Ph. Eur. volumetric standard, secondary reference material for alkalimetry, Supelco®
Supplier: Merck
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
Supplier: Merck
Potassium chloride 3 mol/l (3 N) in aqueous solution, Supelco®
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
HPLC columns, SUPELCOSIL™, LC-18, Supelco®
Supplier: Merck