Residual Solvent - Chloroform Pharmaceutical Secondary Standard Certified Reference Material 1 * 3 items
Catalog # PHR1316-3X1.2M
Supplier: Merck
Residual Solvent - Chloroform Pharmaceutical Secondary Standard Certified Reference Material 1 * 3 items
Catalog # PHR1316-3X1.2M
Supplier: Merck
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
pH buffers, ready to use
Supplier: WTW
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals
Buffer solutions in sachets
Supplier: Mettler - Toledo
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals