Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
6 intake manifold with CN 29/32 valve 1 * 1 items
TELRFB638432
:
Azbil Telstar Technologies
6 intake manifold with CN 29/32 valve 1 * 1 items
TELRFB638432
:
Azbil Telstar Technologies
Frequently Bought Together
Sacrifical body 1 * 1 items
: VWR Collection
Accessories for electronic dispensers, Multipette E3/E3x
: EPPENDORF
Accessories for Research® Plus pipettes
: EPPENDORF
Mains Cable H11 Spare 1 * 1 items
: IKA
Filter protection 5 ml 1 * 50 items
: EPPENDORF
Lithium Polymer Rechargable Battery
: EPPENDORF
Accessories for accu-jet® pro
: Brand
VWR®, Accessories for Mechanical Pipettes
: VWR Collection
Accessories for Titrette®
: Brand
Cover gasket 1 * 1 items
: VWR Collection
BIOTECTOR BASE REAGENT 1 * 20 L
: Hach
Accessories for dispersing elements for homogenisers
: IKA