Order Entry
Ireland
ContactUsLinkComponent
Process2Clean 5 is a highly built neutral pH cleaning agent that can be used as a stand-alone detergent or as an additive to improve the cleaning performance of all VAI cleaning agents. Ideal for use on most all substrate material and effective on m 1 * 1 Bottle
Catalog # VELTPC-5-55G-01
Supplier:  VELTEK ASSOCIATES
Process2Clean 5 is a highly built neutral pH cleaning agent that can be used as a stand-alone detergent or as an additive to improve the cleaning performance of all VAI cleaning agents. Ideal for use on most all substrate material and effective on m 1 * 1 Bottle
Catalog # VELTPC-5-55G-01
Supplier:  VELTEK ASSOCIATES
Frequently Bought Together