Order Entry
Ireland
ContactUsLinkComponent
20x and 60x water immersion system includes 20x apo lwd 0.95 na and 60x plan apo 1.20 na water immersion objectives and dispenser mo dule 1 * 1 items
Catalog # MLDV5076628
Supplier:  Molecular Devices
20x and 60x water immersion system includes 20x apo lwd 0.95 na and 60x plan apo 1.20 na water immersion objectives and dispenser mo dule 1 * 1 items
Catalog # MLDV5076628
Supplier:  Molecular Devices
Frequently Bought Together