Mercury (II) (Mercuric) Nitrate 0.05M (0.1N) CRM, ISO 17034 and ISO 17025 Conservation 24 months. Store under normal laboratory conditions, at temperatures between 15C to 25C 1 * 500 mL
Catalog # CMPLZ14041042
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together