1-Stearoyl-2-15(S)-HETE-sn-glycero-3-PE-d11 1 * 50 µG
Catalog # CAYM27931-50
Supplier: Cayman Chemical
1-Stearoyl-2-15(S)-HETE-sn-glycero-3-PE-d11 1 * 50 µG
Catalog # CAYM27931-50
Supplier: Cayman Chemical
Frequently Bought Together
4-Methyl-2-pentanone ≥97%, TECHNICAL
Supplier: VWR Chemicals
Phenol liquified ≥90% for synthesis
Supplier: Merck
Methyl acetate anhydrous, 99.5 % 1 * 1 L
Supplier: Merck
Ethyl acetate ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
1,4-Dioxane ≥99% for HPLC
Supplier: Thermo Fisher Scientific
Acetonitrile ≥99.9%, CHROMASOLV™, gradient grade for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetonitrile, extra dry 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
Vaseline Oil (USP, BP, Ph. Eur.) pure, pharma grade 1 * 1.000 mL
Supplier: PanReac AppliChem
Chloroform ≥99.5% stabilised
Supplier: Honeywell Chemicals
Water ACS, Deionized
Supplier: Thermo Fisher Scientific
Methanol ≥99.8% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
Dodecyltrimethylammonium bromide, Supelco®
Supplier: Merck
Acetonitrile ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals