Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
500ML BUFFER SOLUTION PH6.00 (PHOSPHATE), NIST STANDARD SOLUTION READY TO USE FOR PH MEASUREMENT 1 * 500 mL
Catalog # MSPP10690404
Supplier:
MARKET SOURCE PART PROCESS
Expand image
500ML BUFFER SOLUTION PH6.00 (PHOSPHATE), NIST STANDARD SOLUTION READY TO USE FOR PH MEASUREMENT 1 * 500 mL
Catalog # MSPP10690404
Supplier:
MARKET SOURCE PART PROCESS
Frequently Bought Together
Supelco® PESTANAL® Organic Reference Standard, Hexazinone
Supplier: Merck
Decitabine, Reference Standard
Supplier: USP
Calibration fluids and oils for refractometers
Supplier: BELLINGHAM STANLEY
Alpha Cyclodextrin, Reference Standard
Supplier: USP
Clopidogrel Diester Analog, Pharmaceutical Analytical Impurities
Supplier: USP
Lindane, Reference Standards
Supplier: USP
Reagents for alkalinity determination using Lovibond® MD photometers
Supplier: Lovibond
Organic reference standard, Aroclor 1260 (PCB-mixture, 60% chlorinated) 1000 µg/ml in isooctane, Supelco®
Supplier: Merck
Supelco® Organic Reference Standard, Acetyleugenol
Supplier: Merck
Alpha Tocopheryl Acid Succinate, Reference Standard
Supplier: USP
Supelco® Organic Reference Standard, Geraniol solution
Supplier: Merck
Pharmaceutical secondary standard, ofloxacin, (CRM), Supelco®
Supplier: Merck
4-ISOPROPYLPHENOL 1 * 50 mg
Supplier: Merck