Order Entry
Ireland
ContactUsLinkComponent
Video-/photo tube A,for M-series, required video objective for: 1/3 inch cameras 0.32x (10445928) 1/2 inch cameras 0.5x (10445929) 2/3 inch cameras 0.63x (10447367 or 10446307) 1 inch cameras 1.0x (10445930 with 11541006) 1 * 1 items
Catalog # LEIM10445925
Supplier:  LEICA MICROSYSTEMS
Video-/photo tube A,for M-series, required video objective for: 1/3 inch cameras 0.32x (10445928) 1/2 inch cameras 0.5x (10445929) 2/3 inch cameras 0.63x (10447367 or 10446307) 1 inch cameras 1.0x (10445930 with 11541006) 1 * 1 items
Catalog # LEIM10445925
Supplier:  LEICA MICROSYSTEMS
Frequently Bought Together