ACQUITY UPLC HSS C18 SB Method Validation Kit, 100Å, 1.8 µm, 3 mm X 100 mm 1 * 3 items
Catalog # WATE186004709
Supplier: WATERS
ACQUITY UPLC HSS C18 SB Method Validation Kit, 100Å, 1.8 µm, 3 mm X 100 mm 1 * 3 items
Catalog # WATE186004709
Supplier: WATERS
Frequently Bought Together
Silica gel, Pore size: 60 Å 0.063-0.200 mm (70-230 mesh ASTM) for column chromatography, Millipore®
Supplier: MERCK PRODUCTION CHEMICALS
Air valves for safetycaps generation 2.0
Supplier: S.C.A.T.
VWR®, Vial Racks
Supplier: VWR Collection
VWR®, Crimp Caps for Crimp Neck Vials ND11
Supplier: VWR Collection
VWR®, Screw Neck Vials ND13
Supplier: VWR Collection
Exhaust filters, V3.0
Supplier: S.C.A.T.
VWR®, Screw Neck Vials ND24
Supplier: VWR Collection
Headspace crimp caps with 20 mm septa
Supplier: Agilent
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Screw Neck Headspace Vials ND18
Supplier: VWR Collection
Florisil® 60-100 mesh for chromatography
Supplier: VWR Chemicals
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection