CORTECS HILIC Method Validation Kit, 90Å, 2.7 µm, 2.1 mm X 30 mm 1 * 3 items
Catalog # WATE186007424
Supplier: WATERS
CORTECS HILIC Method Validation Kit, 90Å, 2.7 µm, 2.1 mm X 30 mm 1 * 3 items
Catalog # WATE186007424
Supplier: WATERS
Frequently Bought Together
Macro cuvettes
Supplier: Agilent
Guard cartridges chiralpak® im-3 10x4mm 3μm 1 * 3 items
Supplier: CHIRAL TECHNOLOGIES
Gold-grade glass screw and crimp top 2 ml vials, level 3 high performance applications, SureSTART™
Supplier: Thermo Fisher Scientific
ISAspher HPLC Columns, C18 BDS
Supplier: ISERA
FRIT BLOCK FLANGE NW300 B8003165 1 * 1 items
Supplier: MERKLINGER PRAZISION WERKZEUG
Supplies for Agilent Instruments 7700 Series ICP-MS
Supplier: Agilent
Avantor® ACE® UltraCore SuperPhenylHexyl, HPLC/UHPLC Columns, 2,5 µm
Supplier: Avantor
250F-LMARK-PE-5.1/0.72LC 250UL SYR (LMK.2620820) 1 * 1 items
Supplier: Trajan Scientific and Medical
Accessories for glass block manifolds, HyperSep™
Supplier: Thermo Fisher Scientific