CORTECS HILIC Method Validation Kit, 90Å, 1.6 µm, 2.1 mm X 30 mm 1 * 3 items
Catalog # WATE186007166
Supplier: WATERS
CORTECS HILIC Method Validation Kit, 90Å, 1.6 µm, 2.1 mm X 30 mm 1 * 3 items
Catalog # WATE186007166
Supplier: WATERS
Frequently Bought Together
Avantor® ACE® C18, HPLC Columns, 5 µm
Supplier: Avantor
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
Modified syringes Microliter™7000 series
Supplier: HAMILTON BONADUZ
Florisil® 60-100 mesh for chromatography
Supplier: VWR Chemicals
VWR® Vial Racks
Supplier: VWR Collection
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
VWR®, Storage Boxes for Sample Vials and Bottles, PP
Supplier: VWR Collection
VWR®, Crimp Caps for Crimp Neck Vials ND11
Supplier: VWR Collection
Avantor® Alltima, HPLC Columns, C18
Supplier: Avantor
VWR®, Snap Cap Vials
Supplier: VWR Collection
Precision screw caps, 18 mm, level 3 high performance applications, SureSTART™
Supplier: Thermo Fisher Scientific
FlashPure Select Silica Cartridges
Supplier: BUCHI
VWR®, Screw Neck Vials ND10
Supplier: VWR Collection