ACQUITY UPLC CSH C18 Method Validation Kit, 130Å, 1.7 µm, 2.1 mm X 100 mm 1 * 3 items
Catalog # WATE186005572
Supplier: WATERS
ACQUITY UPLC CSH C18 Method Validation Kit, 130Å, 1.7 µm, 2.1 mm X 100 mm 1 * 3 items
Catalog # WATE186005572
Supplier: WATERS
Frequently Bought Together
Extraction unit and drying attachment, LiChrolut®
Supplier: Merck
FlashPure Select Silica Cartridges
Supplier: BUCHI
Glass snap top vials, 2 ml, level 2 high-throughput applications, SureSTART™
Supplier: Thermo Fisher Scientific
Capillary columns, OPTIMA® 5
Supplier: MACHEREY-NAGEL
HPLC and GC certified screw vial and cap kits, level 3 high performance applications, SureSTART™
Supplier: Thermo Fisher Scientific
Chromatography Tubing, Stainless Steel
Supplier: Avantor
ISAspher HPLC Columns, C8
Supplier: ISERA
ISAspher HPLC Columns, C18 BDS
Supplier: ISERA
Supplies for Agilent Instruments 7700 Series ICP-MS
Supplier: Agilent
HPLC columns, NUCLEOSIL® SA
Supplier: MACHEREY-NAGEL
1/32 FS adapters 1x 5St. 1 * 5 items
Supplier: MACHEREY-NAGEL