ACQUITY UPLC Peptide BEH C18 Method Validation Kit, 130Å, 1.7 µm, 2.1 mm X 100 mm, 1K - 15K 1 * 3 items
Catalog # WATE186004896
Supplier: WATERS
ACQUITY UPLC Peptide BEH C18 Method Validation Kit, 130Å, 1.7 µm, 2.1 mm X 100 mm, 1K - 15K 1 * 3 items
Catalog # WATE186004896
Supplier: WATERS
Frequently Bought Together
SafetyWasteCaps
Supplier: S.C.A.T.
VWR®, Screw Caps for Short Thread Vials ND9
Supplier: VWR Collection
Avantor® ACE® Excel® SuperC18, HPLC/UHPLC Columns, Analytical, 3 µm
Supplier: Avantor
Avantor® ACE®, Guard Cartridge Holders
Supplier: Avantor
Exhaust filters, V3.0
Supplier: S.C.A.T.
VWR®, Screw Neck Vials ND24
Supplier: VWR Collection
Glass capillaries and graduated micropipettes, disposable
Supplier: CAMAG
Avantor® ACE® C18, HPLC Columns, 5 µm
Supplier: Avantor
Accessories for TLC sprayer
Supplier: Merck
Accessories for connection system GLS 80
Supplier: DWK Life Sciences
Air valves for safetycaps generation 2.0
Supplier: S.C.A.T.
VWR®, Storage Boxes for Sample Vials and Bottles, PP
Supplier: VWR Collection
Avantor® Partisil SAX, HPLC Columns
Supplier: Avantor
Canisters, GL 45, b.safe
Supplier: Bohlender
Gold standard syringes for autosampler
Supplier: Agilent
Bonded screw caps for screw neck vials, N 15 (15-425)
Supplier: MACHEREY-NAGEL
Glass crimp top headspace vials, 20 ml, level 2 high-throughput applications, SureSTART™
Supplier: Thermo Fisher Scientific