CORTECS HILIC Method Validation Kit, 90Å, 1.6 µm, 2.1 mm X 100 mm 1 * 3 items
WATE186007169
: WATERS
CORTECS HILIC Method Validation Kit, 90Å, 1.6 µm, 2.1 mm X 100 mm 1 * 3 items
WATE186007169
: WATERS
Frequently Bought Together
Florisil® 60-100 mesh for chromatography
: VWR Chemicals
VWR®, Crimp Caps for Crimp Neck Vials ND11
: VWR Collection
SafetyCaps
: S.C.A.T.
VWR®, Screw Neck Vials ND8
: VWR Collection
Exhaust filters, V3.0
: S.C.A.T.
VWR®, Screw Neck Vials ND24
: VWR Collection
VWR®, Short Thread Vials ND9
: VWR Collection
VWR®, Screw Neck Headspace Vials ND18
: VWR Collection
VWR®, Screw Caps for Short Thread Vials ND9
: VWR Collection
Screw neck vials
: WATERS
VWR®, Crimp Caps for Crimp Neck Headspace Vials ND20
: VWR Collection
Air valves for safetycaps generation 2.0
: S.C.A.T.
Septa
: METROHM
Certified headspace crimp top vials
: Agilent
Inlet liners
: Agilent
Screw caps for certified screw top vials
: Agilent