ACQUITY UPLC BEH Amide Method Validation Kit, 130Å, 1.7 µm, 2.1 mm X 50 mm 1 * 3 items
Catalog # WATE186004807
Supplier: WATERS
ACQUITY UPLC BEH Amide Method Validation Kit, 130Å, 1.7 µm, 2.1 mm X 50 mm 1 * 3 items
Catalog # WATE186004807
Supplier: WATERS
Frequently Bought Together
Spare needles for 1 to 2,5 ml syringes
Supplier: Trajan Scientific and Medical
Gold-grade glass screw and crimp top 2 ml vials, level 3 high performance applications, SureSTART™
Supplier: Thermo Fisher Scientific
250F-LMARK-PE-5.1/0.72LC 250UL SYR (LMK.2620820) 1 * 1 items
Supplier: Trajan Scientific and Medical
HPLC columns, Waters Spherisorb®
Supplier: WATERS
Avantor® ACE® UltraCore SuperPhenylHexyl, HPLC/UHPLC Columns, 2,5 µm
Supplier: Avantor
ISAspher HPLC Columns, C18 AQ
Supplier: ISERA
Horizontal developing chambers
Supplier: CAMAG
XELA UHPLC Columns, HILIC-Diol
Supplier: ISERA
Accessories for HPLC columns Acclaim™
Supplier: Thermo Fisher Scientific