Order Entry
Ireland
ContactUsLinkComponent
XSelect CSH C18 Method Validation Kit, 130Å, 3.5 µm, 4.6 mm X 100 mm 1 * 3 items
Catalog # WATE186005541
Supplier:  WATERS
XSelect CSH C18 Method Validation Kit, 130Å, 3.5 µm, 4.6 mm X 100 mm 1 * 3 items
Catalog # WATE186005541
Supplier:  WATERS
Frequently Bought Together