XBridge BEH Shield RP18 Method Validation Kit, 130Å, 2.5 µm, 4.6 mm X 50 mm 1 * 3 items
Catalog # WATE186006213
Supplier: WATERS
XBridge BEH Shield RP18 Method Validation Kit, 130Å, 2.5 µm, 4.6 mm X 50 mm 1 * 3 items
Catalog # WATE186006213
Supplier: WATERS
Frequently Bought Together
Safety canisters
Supplier: S.C.A.T.
VWR®, Screw Caps for Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Screw Neck Headspace Vials ND18
Supplier: VWR Collection
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
VWR®, Crimp Caps for Crimp Neck Headspace Vials ND20
Supplier: VWR Collection
Air valves for safetycaps generation 2.0
Supplier: S.C.A.T.
VWR®, Screw Neck Vials ND13
Supplier: VWR Collection
EPA screw neck vials, N 24
Supplier: MACHEREY-NAGEL
Headspace crimp caps with 20 mm septa
Supplier: Agilent
VWR®, Snap Cap Vials
Supplier: VWR Collection
Exhaust filters, V3.0
Supplier: S.C.A.T.
Accessories for GC/MS
Supplier: Agilent
Inlet septa
Supplier: Agilent