CORTECS HILIC Method Validation Kit, 90Å, 2.7 µm, 4.6 mm X 75 mm 1 * 3 items
WATE186007436
: WATERS
CORTECS HILIC Method Validation Kit, 90Å, 2.7 µm, 4.6 mm X 75 mm 1 * 3 items
WATE186007436
: WATERS
Frequently Bought Together
GC capillary column connectors, Swagelok®
: PerkinElmer
Avantor® Hichrom, HPLC Columns, RPB
: Avantor
Jars, wide-mouth, with open top cap
: EP SCIENTIFIC
ISAspher HPLC Columns, C8
: ISERA
HPLC columns, Accucore™ RP-MS
: Thermo Fisher Scientific
HPLC columns, AquaSil™ C18
: Thermo Fisher Scientific
High purity brass gas regulators
: Thermo Fisher Scientific
Micro extraction by packed sorbent (MEPS™)
: Trajan Scientific and Medical
One-piece fittings with integrated ferrules
: S.C.A.T.
1290 valve head capillaries
: Agilent