Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
XSelect Peptide CSH C18 Column Kit, 130Å, 2.5 µm, 2.1 mm X 150 mm, 1K - 15K 1 * 1 items
WATE176003071
:
WATERS
XSelect Peptide CSH C18 Column Kit, 130Å, 2.5 µm, 2.1 mm X 150 mm, 1K - 15K 1 * 1 items
WATE176003071
:
WATERS
Frequently Bought Together
Vydac 214TP C4, HPLC columns
: Avantor
Avantor® ACE® C18, HPLC Columns, 5 µm
: Avantor
TLC plates, silica gel 60, unmodified layers, plastic sheets
: Merck
Avantor® Ultrasphere®, HPLC Columns
: Avantor
Screw neck vials, N 8
: MACHEREY-NAGEL
SPE columns, hydrocarbons analysis, CHROMABOND® Na₂SO₄/Florisil®
: MACHEREY-NAGEL
Syringes for thin layer chromatography
: HAMILTON BONADUZ
VWR®, Manual Crimper and Decapper
: VWR Collection
Supplies for pumps
: Agilent
Cellulose DS-0, powder suitable for thin layer chromatography (TLC), Supelco®
: Merck
CORTECS Phenyl Method Validation Kit, 90Å, 2.7 µm, 2.1 mm X 100 mm 1 * 3 items
: WATERS
OASIS WAX CARTRIDGE 6 CC/150 MG 1 * 30 items
: WATERS
LC vials and closures
: Agilent