ACQUITY Premier Peptide CSH C18, 130Å, 1.7 µm 2.1 x 150 mm MVK 1 * 3 items
Catalog # WATE186009653
Supplier: WATERS
ACQUITY Premier Peptide CSH C18, 130Å, 1.7 µm 2.1 x 150 mm MVK 1 * 3 items
Catalog # WATE186009653
Supplier: WATERS
Frequently Bought Together
VWR® ACE® Excel® C18 HPLC/UHPLC Columns, Analytical, 2 µm
Supplier: Avantor
GC COLUMN HI-1 30M 0.32MM 1.00UM 5IN CAG 1 * 1 items
Supplier: Avantor
TLC plates, silica gel 60, unmodified layers, aluminium sheets, ALUGRAM® Xtra SILG
Supplier: MACHEREY-NAGEL
Cation exchange HPLC columns, PRP-X800
Supplier: HAMILTON BONADUZ
Seals and septa for headspace crimp seal vials, 20 mm
Supplier: DWK Life Sciences
Gas cartridge filters, Super Clean™
Supplier: Thermo Fisher Scientific
L-MARK Gold Syringe 100UL Fixed Needle, Black PTFE, 51mm Length, 22s Gauge, LC Tip PK6 1 * 6 items
Supplier: Trajan Scientific and Medical
Avantor® ACE® Silica, HPLC Columns, 5 µm
Supplier: Avantor
HPLC columns, Hypersil™ BDS C18
Supplier: Thermo Fisher Scientific
Tubing, flanged, PTFE, BOLA
Supplier: Bohlender
VWR®, HPTLC Glass/Aluminium Plates, Silica, Modified Layers
Supplier: VWR Collection