ACQUITY Premier Peptide BEH C18, 300Å, 1.7 µm 2.1 x 100 mm MVK 1 * 3 items
Catalog # WATE186009646
Supplier: WATERS
ACQUITY Premier Peptide BEH C18, 300Å, 1.7 µm 2.1 x 100 mm MVK 1 * 3 items
Catalog # WATE186009646
Supplier: WATERS
Frequently Bought Together
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Screw Caps for Short Thread Vials ND9
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR®, Screw Neck Vials ND8
Supplier: VWR Collection
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Snap Cap Vials
Supplier: VWR Collection
VWR®, Screw Neck Vials ND24
Supplier: VWR Collection
VWR®, Inserts for Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals