Order Entry
Ireland
ContactUsLinkComponent
XBridge BEH HILIC Column, 130Å, 2.5 µm, 3 mm X 100 mm 1 * 3 items
Catalog # WATE176002585
Supplier:  WATERS
XBridge BEH HILIC Column, 130Å, 2.5 µm, 3 mm X 100 mm 1 * 3 items
Catalog # WATE176002585
Supplier:  WATERS
Frequently Bought Together