Order Entry
Ireland
ContactUsLinkComponent
Atlantis Silica HILIC OBD Prep Column, 100Å, 5 µm, 19 mm X 100 mm 1 * 1 items
Catalog # WATE186003958
Supplier:  WATERS
Atlantis Silica HILIC OBD Prep Column, 100Å, 5 µm, 19 mm X 100 mm 1 * 1 items
Catalog # WATE186003958
Supplier:  WATERS
Frequently Bought Together