XBridge BEH C8 OBD Prep Column, 130Å, 5 µm, 19 mm X 50 mm 1 * 1 items
Catalog # WATE186002993
Supplier: WATERS
XBridge BEH C8 OBD Prep Column, 130Å, 5 µm, 19 mm X 50 mm 1 * 1 items
Catalog # WATE186002993
Supplier: WATERS
Frequently Bought Together
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Screw Neck Headspace Vials ND18
Supplier: VWR Collection
VWR®, Screw Caps for Screw Neck Vials ND13
Supplier: VWR Collection
VWR®, Screw Neck Vials ND13
Supplier: VWR Collection
Vial racks
Supplier: MACHEREY-NAGEL
VWR®, Screw Caps for Short Thread Vials ND9
Supplier: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Exhaust filters, V3.0
Supplier: S.C.A.T.
VWR®, Screw Neck Vials ND24
Supplier: VWR Collection
Florisil® 60-100 mesh for chromatography
Supplier: VWR Chemicals
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
VWR®, Inserts for Short Thread Vials ND9
Supplier: VWR Collection