Order Entry
Ireland
ContactUsLinkComponent
Atlantis T3 OBD Prep Column, 100Å, 5 µm, 19 mm X 50 mm 1 * 1 items
  WATE186003696
 :  WATERS
Atlantis T3 OBD Prep Column, 100Å, 5 µm, 19 mm X 50 mm 1 * 1 items
  WATE186003696
 :  WATERS
Frequently Bought Together