XSelect CSH C18 OBD Prep Column, 130Å, 5 µm, 10 mm X 250 mm 1 * 1 items
Catalog # WATE186008239
Supplier: WATERS
XSelect CSH C18 OBD Prep Column, 130Å, 5 µm, 10 mm X 250 mm 1 * 1 items
Catalog # WATE186008239
Supplier: WATERS
Frequently Bought Together
VWR®, Screw Caps for Short Thread Vials ND9
Supplier: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Screw Neck Headspace Vials ND18
Supplier: VWR Collection
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR®, Screw Neck Vials ND13
Supplier: VWR Collection
Exhaust filters, V3.0
Supplier: S.C.A.T.
VWR®, Screw Neck Vials ND24
Supplier: VWR Collection
VWR®, Snap Cap Vials
Supplier: VWR Collection