Delta-Pak C4 Radial-Pak Column, 300Å, 15 µm, 40 mm X 100 mm, 1K - 30K 1 * 1 items
Catalog # WATEWAT037700
Supplier: WATERS
Delta-Pak C4 Radial-Pak Column, 300Å, 15 µm, 40 mm X 100 mm, 1K - 30K 1 * 1 items
Catalog # WATEWAT037700
Supplier: WATERS
Frequently Bought Together
Avantor® ACE® Excel® SuperC18, HPLC/UHPLC Columns, Analytical, 5 µm
Supplier: Avantor
Avantor® ACE® C8, HPLC Columns, 5 µm
Supplier: Avantor
Accessories for connection system GLS 80
Supplier: DWK Life Sciences
20Mm Borate Buffer Ph9.3 100Ml 1 * 100 mL
Supplier: Agilent
Pre-packed columns, 50 and 200 ml, SkillPak™
Supplier: TOSOH Bioscience
Flash chromatography cartridges, CHROMABOND® Flash RS
Supplier: MACHEREY-NAGEL
Syringes, Neuros™
Supplier: HAMILTON BONADUZ
LC vials and closures
Supplier: Agilent
ISAspher HPLC Columns, Diol, HILIC
Supplier: ISERA
HPLC columns, NUCLEOSIL® C18
Supplier: MACHEREY-NAGEL
HPLC columns, SUPELCOSIL™, LC-8-DB, Supelco®
Supplier: Merck
Canisters, S60/61, with level indicator, b.safe
Supplier: Bohlender
HPLC columns for carbohydrate analysis, Hi-Plex
Supplier: VARIAN