XTerra Shield RP18 Intelligent Speed (IS) Column, 125Å, 5 µm, 2.1 mm X 20 mm 1 * 1 items
Catalog # WATE186001982
Supplier: WATERS
XTerra Shield RP18 Intelligent Speed (IS) Column, 125Å, 5 µm, 2.1 mm X 20 mm 1 * 1 items
Catalog # WATE186001982
Supplier: WATERS
Frequently Bought Together
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
VWR® Air Valve
Supplier: VWR Collection
Vial racks
Supplier: MACHEREY-NAGEL
Exhaust filters, V3.0
Supplier: S.C.A.T.
VWR®, Screw Neck Vials ND24
Supplier: VWR Collection
VWR®, Snap Cap Vials
Supplier: VWR Collection
Silica gel 60, irregular 40 - 63 µm for flash chromatography
Supplier: VWR Chemicals
VWR®, Screw Caps for Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Manual Crimper and Decapper
Supplier: VWR Collection
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
Florisil® 60-100 mesh for chromatography
Supplier: VWR Chemicals
VWR®, Inserts for Short Thread Vials ND9
Supplier: VWR Collection
Air valves for safetycaps generation 2.0
Supplier: S.C.A.T.
VWR®, TLC Aluminium Plates, Silica, Unmodified Layers
Supplier: VWR Collection
VWR®, Screw Neck Vials ND13
Supplier: VWR Collection