Styragel HMW 6E, mixed bed, 20 µm, 4.6 mm X 300 mm in DMF, 5K - 10M 1 * 1 items
Catalog # WATEWAT046825
Supplier: WATERS
Styragel HMW 6E, mixed bed, 20 µm, 4.6 mm X 300 mm in DMF, 5K - 10M 1 * 1 items
Catalog # WATEWAT046825
Supplier: WATERS
Frequently Bought Together
Collecting trays, b.safe
Supplier: Bohlender
Florisil® 60-100 mesh for chromatography
Supplier: VWR Chemicals
VWR®, Manual Crimper and Decapper
Supplier: VWR Collection
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Crimp Caps for Crimp Neck Vials ND11
Supplier: VWR Collection
VWR®, Screw Caps for Screw Neck Vials ND10
Supplier: VWR Collection
VWR®, Storage Boxes for Sample Vials and Bottles, PP
Supplier: VWR Collection
VWR®, Crimp Caps for Crimp Neck Headspace Vials ND20
Supplier: VWR Collection
VWR®, Screw Neck Vials ND24
Supplier: VWR Collection
VWR®, Snap Cap Vials
Supplier: VWR Collection
Headspace crimp caps with 20 mm septa
Supplier: Agilent
Flat bottom chambers
Supplier: CAMAG
N 20 stopper Bromobutyl, gray null
Supplier: MACHEREY-NAGEL
TSKgel® gel filtration columns, PW-type
Supplier: TOSOH Bioscience