Order Entry
Ireland
ContactUsLinkComponent
Spherisorb Phenyl Column, 80Å, 3 µm, 4.6 mm X 50 mm 1 * 1 items
  WATEPSS833711
 :  WATERS
Spherisorb Phenyl Column, 80Å, 3 µm, 4.6 mm X 50 mm 1 * 1 items
  WATEPSS833711
 :  WATERS
Frequently Bought Together