Delta-Pak C18 Radial-Pak Guard, 300Å, 15 µm, 25 mm X 10 mm, 1K - 30K 1 * 2 items
Catalog # WATEWAT038522
Supplier: WATERS
Delta-Pak C18 Radial-Pak Guard, 300Å, 15 µm, 25 mm X 10 mm, 1K - 30K 1 * 2 items
Catalog # WATEWAT038522
Supplier: WATERS
Frequently Bought Together
VWR® Air Valve
Supplier: VWR Collection
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Screw Neck Headspace Vials ND18
Supplier: VWR Collection
VWR®, Screw Neck Vials ND8
Supplier: VWR Collection
VWR®, Screw Caps for Short Thread Vials ND9
Supplier: VWR Collection
VWR®, TLC Aluminium Plates, Silica, Unmodified Layers
Supplier: VWR Collection
Air valves for safetycaps generation 2.0
Supplier: S.C.A.T.
VWR®, Inserts for Short Thread Vials ND9
Supplier: VWR Collection
VWR® Standard Micro Syringes
Supplier: VWR Collection
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
VWR®, Crimp Caps for Crimp Neck Vials ND11
Supplier: VWR Collection
VWR®, Screw Neck Vials ND13
Supplier: VWR Collection
EPA screw neck vials, N 24
Supplier: MACHEREY-NAGEL
Exhaust filters, V3.0
Supplier: S.C.A.T.
VWR®, Snap Cap Vials
Supplier: VWR Collection
VWR®, Screw Neck Vials ND24
Supplier: VWR Collection