ACQUITY UPLC CSH Phenyl-Hexyl Column, 130Å, 1.7 µm, 3 mm X 150 mm 1 * 1 items
Catalog # WATE186005412
Supplier: WATERS
ACQUITY UPLC CSH Phenyl-Hexyl Column, 130Å, 1.7 µm, 3 mm X 150 mm 1 * 1 items
Catalog # WATE186005412
Supplier: WATERS
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Screw Neck Headspace Vials ND18
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Crimp Caps for Crimp Neck Vials ND11
Supplier: VWR Collection
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
Florisil® 60-100 mesh for chromatography
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR®, Snap Cap Vials
Supplier: VWR Collection
Exhaust filters, V3.0
Supplier: S.C.A.T.
Convenience kits for inlet systems, QuickPick
Supplier: Agilent