Order Entry
Ireland
ContactUsLinkComponent
Atlantis Silica HILIC Column, 100Å, 3 µm, 3 mm X 100 mm 1 * 1 items
Catalog # WATE186002019
Supplier:  WATERS
Atlantis Silica HILIC Column, 100Å, 3 µm, 3 mm X 100 mm 1 * 1 items
Catalog # WATE186002019
Supplier:  WATERS
Frequently Bought Together