XSelect CSH C18 OBD Prep Column, 130Å, 5 µm, 19 mm X 250 mm 1 * 1 items
Catalog # WATE186005492
Supplier: WATERS
XSelect CSH C18 OBD Prep Column, 130Å, 5 µm, 19 mm X 250 mm 1 * 1 items
Catalog # WATE186005492
Supplier: WATERS
Frequently Bought Together
VWR®, Crimp Caps for Crimp Neck Headspace Vials ND20
Supplier: VWR Collection
VWR®, Screw Neck Vials ND13
Supplier: VWR Collection
VWR®, Storage Boxes for Sample Vials and Bottles, PP
Supplier: VWR Collection
Exhaust filters, V3.0
Supplier: S.C.A.T.
EPA screw neck vials, N 24
Supplier: MACHEREY-NAGEL
VWR®, Snap Cap Vials
Supplier: VWR Collection
VWR®, Inserts for Screw Neck Vials ND8
Supplier: VWR Collection
VWR®, Screw Neck Vials ND24
Supplier: VWR Collection
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
Crimp neck vials, N 20
Supplier: MACHEREY-NAGEL
Florisil® 60-100 mesh for chromatography
Supplier: VWR Chemicals
FlashPure Select Silica Cartridges
Supplier: BUCHI
Certified headspace crimp top vials
Supplier: Agilent
Gold standard syringes for autosampler
Supplier: Agilent
Screw caps for glass screw top vials, 9 mm, level 1 everyday analysis, SureSTART™
Supplier: Thermo Fisher Scientific