Order Entry
Ireland
ContactUsLinkComponent
Symmetry C18 Column, 100Å, 3.5 µm, 2.1 mm X 30 mm 1 * 1 items
Catalog # WATEWAT058973
Supplier:  WATERS
Symmetry C18 Column, 100Å, 3.5 µm, 2.1 mm X 30 mm 1 * 1 items
Catalog # WATEWAT058973
Supplier:  WATERS
Frequently Bought Together