Symmetry C18 Column, 300Å, 5 µm, 3.9 mm X 150 mm 1 * 1 items
Catalog # WATEWAT106154
Supplier: WATERS
Symmetry C18 Column, 300Å, 5 µm, 3.9 mm X 150 mm 1 * 1 items
Catalog # WATEWAT106154
Supplier: WATERS
Frequently Bought Together
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
Bulb pipettes (one mark), BLAUBRAND®, class AS
Supplier: Brand
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Screw Caps for Short Thread Vials ND9
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
VWR®, Snap Cap Vials
Supplier: VWR Collection
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals