XTerra Shield RP8 Column, 125Å, 3.5 µm, 3 mm X 100 mm 1 * 1 items
Catalog # WATE186000421
Supplier: WATERS
XTerra Shield RP8 Column, 125Å, 3.5 µm, 3 mm X 100 mm 1 * 1 items
Catalog # WATE186000421
Supplier: WATERS
Frequently Bought Together
VWR®, Manual Crimper and Decapper
Supplier: VWR Collection
VWR®, Screw Caps for Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Crimp Caps for Crimp Neck Headspace Vials ND20
Supplier: VWR Collection
VWR®, Septa GL 45
Supplier: VWR Collection
VWR®, Screw Caps for Screw Neck Vials ND24
Supplier: VWR Collection
Vial racks
Supplier: MACHEREY-NAGEL
VWR®, Screw Neck Vials ND24
Supplier: VWR Collection
VWR®, Snap Cap Vials
Supplier: VWR Collection
Crimp caps for crimp neck vials, N 20
Supplier: MACHEREY-NAGEL
Screw neck vials, N 9
Supplier: MACHEREY-NAGEL
VWR®, Snap Ring Vials ND11
Supplier: VWR Collection
Avantor® ACE® C18, HPLC Columns, 5 µm
Supplier: Avantor
VWR®, Screw Neck Vials ND13
Supplier: VWR Collection
VWR®, Storage Boxes for Sample Vials and Bottles, PP
Supplier: VWR Collection
Autosampler syringes for 7673/7683 autosamplers
Supplier: Agilent