ACQUITY UPLC Peptide HSS T3 Column, 100Å, 1.8 µm, 2.1 mm X 150 mm, 1K - 15K 1 * 1 items
Catalog # WATE186008756
Supplier: WATERS
ACQUITY UPLC Peptide HSS T3 Column, 100Å, 1.8 µm, 2.1 mm X 150 mm, 1K - 15K 1 * 1 items
Catalog # WATE186008756
Supplier: WATERS
Frequently Bought Together
FlashPure Select Silica Cartridges
Supplier: BUCHI
Capillary column ferrules for gas chromatography
Supplier: Agilent
ISAspher HPLC Columns, Amino
Supplier: ISERA
Accessories for optima® 4X00
Supplier: PerkinElmer
SunFire C18 Column, 100Å, 5µm, 4.6mm x 250mm 1 * 1 items
Supplier: WATERS
GC injection port septa
Supplier: MACHEREY-NAGEL
Siliconising Fluid, hydrocarbon soluble
Supplier: Thermo Fisher Scientific
ISAspher HPLC Columns, C18
Supplier: ISERA
CORTECS Phenyl Column, 90Å, 1.6 µm, 3 mm X 150 mm 1 * 1 items
Supplier: WATERS
CORTECS HILIC Column, 90Å, 2.7 µm, 4.6 mm X 30 mm 1 * 1 items
Supplier: WATERS
TSKgel® gel filtration columns, PW-type
Supplier: TOSOH Bioscience