XBridge BEH HILIC Column, 130Å, 3.5 µm, 3 mm X 100 mm 1 * 1 items
Catalog # WATE186004436
Supplier: WATERS
XBridge BEH HILIC Column, 130Å, 3.5 µm, 3 mm X 100 mm 1 * 1 items
Catalog # WATE186004436
Supplier: WATERS
Frequently Bought Together
VWR®, Screw Neck Vials ND8
Supplier: VWR Collection
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Screw Caps for Short Thread Vials ND9
Supplier: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Screw Neck Vials ND24
Supplier: VWR Collection
Exhaust filters, V3.0
Supplier: S.C.A.T.
Silica gel 60, irregular 40 - 63 µm for flash chromatography
Supplier: VWR Chemicals
VWR®, Inserts for Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Crimp Caps for Crimp Neck Headspace Vials ND20
Supplier: VWR Collection
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
Screw caps for certified screw top vials
Supplier: Agilent