Order Entry
Ireland
ContactUsLinkComponent
XBridge BEH HILIC Column, 130Å, 3.5 µm, 2.1 mm X 150 mm 1 * 1 items
Catalog # WATE186004434
Supplier:  WATERS
XBridge BEH HILIC Column, 130Å, 3.5 µm, 2.1 mm X 150 mm 1 * 1 items
Catalog # WATE186004434
Supplier:  WATERS
Frequently Bought Together