CORTECS T3 Column, 120Å, 2.7 µm, 4.6 mm X 75 mm 1 * 1 items
Catalog # WATE186008493
Supplier: WATERS
CORTECS T3 Column, 120Å, 2.7 µm, 4.6 mm X 75 mm 1 * 1 items
Catalog # WATE186008493
Supplier: WATERS
Frequently Bought Together
Safety canisters
Supplier: S.C.A.T.
Avantor® ACE®, Guard Cartridge Holders
Supplier: Avantor
VWR®, Screw Neck Headspace Vials ND18
Supplier: VWR Collection
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
VWR® Standard Micro Syringes
Supplier: VWR Collection
Silica gel, Pore size: 60 Å 0.063-0.200 mm (70-230 mesh ASTM) for column chromatography, Millipore®
Supplier: MERCK PRODUCTION CHEMICALS
Silica gel 60, irregular 40 - 63 µm for flash chromatography
Supplier: VWR Chemicals
EPA screw neck vials, N 24
Supplier: MACHEREY-NAGEL
VWR®, Snap Cap Vials
Supplier: VWR Collection
VWR®, Screw Neck Vials ND24
Supplier: VWR Collection
VWR®, Screw Caps for Short Thread Vials ND9
Supplier: VWR Collection
Screw caps for certified screw top vials
Supplier: Agilent