XBridge BEH HILIC Column, 130Å, 3.5 µm, 4.6 mm X 30 mm 1 * 1 items
Catalog # WATE186004438
Supplier: WATERS
XBridge BEH HILIC Column, 130Å, 3.5 µm, 4.6 mm X 30 mm 1 * 1 items
Catalog # WATE186004438
Supplier: WATERS
Frequently Bought Together
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
VWR®, Screw Neck Vials ND13
Supplier: VWR Collection
Exhaust filters, V3.0
Supplier: S.C.A.T.
VWR®, Snap Cap Vials
Supplier: VWR Collection
VWR®, Screw Neck Vials ND24
Supplier: VWR Collection
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
Glass crimp top headspace vials, 20 ml, level 2 high-throughput applications, SureSTART™
Supplier: Thermo Fisher Scientific
Glass crimp top headspace vials, 20 ml, level 2 high-throughput applications, SureSTART™
Supplier: Thermo Fisher Scientific
Screw caps for certified screw top vials
Supplier: Agilent