Delta-Pak C4 Radial-Pak Guard, 100Å, 15 µm, 25 mm X 10 mm, 1K - 15K 1 * 2 items
Catalog # WATEWAT038524
Supplier: WATERS
Delta-Pak C4 Radial-Pak Guard, 100Å, 15 µm, 25 mm X 10 mm, 1K - 15K 1 * 2 items
Catalog # WATEWAT038524
Supplier: WATERS
Frequently Bought Together
VWR®, Screw Caps for Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Screw Neck Headspace Vials ND18
Supplier: VWR Collection
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
VWR®, TLC Aluminium Plates, Silica, Unmodified Layers
Supplier: VWR Collection
Air valves for safetycaps generation 2.0
Supplier: S.C.A.T.
Silica gel, Pore size: 60 Å 0.063-0.200 mm (70-230 mesh ASTM) for column chromatography, Millipore®
Supplier: MERCK PRODUCTION CHEMICALS
VWR®, Screw Caps for Screw Neck Vials ND24
Supplier: VWR Collection
Screw neck vials, N 9
Supplier: MACHEREY-NAGEL
Exhaust filters, V3.0
Supplier: S.C.A.T.
EPA screw neck vials, N 24
Supplier: MACHEREY-NAGEL
VWR®, Screw Neck Vials ND24
Supplier: VWR Collection
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
VWR®, Crimp Caps for Crimp Neck Vials ND11
Supplier: VWR Collection
VWR®, Screw Neck Vials ND13
Supplier: VWR Collection
VWR®, Storage Boxes for Sample Vials and Bottles, PP
Supplier: VWR Collection