XBridge Premier Protein BEH C4 300 Å 2.5 µm, 4.6 x 150 mm 1 * 1 items
Catalog # WATE186010334
Supplier: WATERS
XBridge Premier Protein BEH C4 300 Å 2.5 µm, 4.6 x 150 mm 1 * 1 items
Catalog # WATE186010334
Supplier: WATERS
Frequently Bought Together
Avantor® ACE® C18-Amide, HPLC/UHPLC Columns, 5 µm
Supplier: Avantor
Replacement needles for syringes, Neuros™
Supplier: HAMILTON BONADUZ
HPLC columns, SeQuant® ZIC®-pHILIC
Supplier: Merck
HPLC columns, MetaCarb
Supplier: VARIAN
SUNFIRE GUARD COLUMN 1 * 1 items
Supplier: WATERS
Supplies for autosamplers, ASX-500 series
Supplier: Agilent
ISAspher HPLC Columns, Cyano
Supplier: ISERA
Spare parts for CE systems
Supplier: Agilent
ICP multi-element standards
Supplier: Agilent
Closures for headspace vials
Supplier: PerkinElmer
Microlab 600 Bubble Free Prime (BFP) syringes
Supplier: HAMILTON BONADUZ