Order Entry
Ireland
ContactUsLinkComponent
Atlantis Premier BEH C18 AX VanGuard FIT Column, 5 µm, 2.1 x 100 mm 1 * 1 items
Catalog # WATE186009405
Supplier:  WATERS
Atlantis Premier BEH C18 AX VanGuard FIT Column, 5 µm, 2.1 x 100 mm 1 * 1 items
Catalog # WATE186009405
Supplier:  WATERS
Frequently Bought Together