SunFire C18 OBD Prep Column, 100Å, 10 µm, 10 mm X 50 mm 1 * 1 items
Catalog # WATE186008208
Supplier: WATERS
SunFire C18 OBD Prep Column, 100Å, 10 µm, 10 mm X 50 mm 1 * 1 items
Catalog # WATE186008208
Supplier: WATERS
Frequently Bought Together
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
Avantor® ACE® C18-PFP, HPLC Columns, 3 µm
Supplier: Avantor
Air valves for safetycaps generation 2.0
Supplier: S.C.A.T.
VWR®, Screw Neck Vials ND8
Supplier: VWR Collection
VWR®, Vial Racks
Supplier: VWR Collection
VWR®, Screw Caps for Screw Neck Vials ND24
Supplier: VWR Collection
VWR®, Septa GL 45
Supplier: VWR Collection
HPLC column accessories
Supplier: Merck
VWR®, Screw Caps for Screw Neck Vials ND13
Supplier: VWR Collection
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Screw Neck Headspace Vials ND18
Supplier: VWR Collection
VWR®, Snap Cap Vials
Supplier: VWR Collection
Exhaust filters, V3.0
Supplier: S.C.A.T.
VWR®, Screw Neck Vials ND13
Supplier: VWR Collection
Inlet septa
Supplier: Agilent