XBridge Peptide BEH C18 OBD Prep Column, 300Å, 5 µm, 10 mm X 50 mm, 1K - 15K 1 * 1 items
Catalog # WATE186008190
Supplier: WATERS
XBridge Peptide BEH C18 OBD Prep Column, 300Å, 5 µm, 10 mm X 50 mm, 1K - 15K 1 * 1 items
Catalog # WATE186008190
Supplier: WATERS
Frequently Bought Together
Safety canisters
Supplier: S.C.A.T.
VWR® ACE® Excel® C18 HPLC/UHPLC Columns, Analytical, 2 µm
Supplier: Avantor
Accessories for flat bottom chambers
Supplier: CAMAG
Crimp Caps for Crimp Neck Vials, N 11
Supplier: MACHEREY-NAGEL
VWR®, Snap Cap Vials
Supplier: VWR Collection
VWR®, Screw Neck Vials ND24
Supplier: VWR Collection
Collecting trays, b.safe
Supplier: Bohlender
Accessories for connection system GLS 80
Supplier: DWK Life Sciences
Solid phase extraction cartridges, Bond Elut™ SI
Supplier: VARIAN
Spare parts for CE systems
Supplier: Agilent
Precision screw caps, 18 mm, level 3 high performance applications, SureSTART™
Supplier: Thermo Fisher Scientific
BOND ELUT JR-NH2 500MG 1 * 100 items
Supplier: VARIAN
Supplies for Agilent Instruments 7700 Series ICP-MS
Supplier: Agilent
SPME fiber assemblies, Supelco®
Supplier: Merck
Caps for LC vials
Supplier: Agilent
Glass crimp top headspace vials, 20 ml, level 1 everyday analysis, SureSTART™
Supplier: Thermo Fisher Scientific