Order Entry
Ireland
ContactUsLinkComponent
XSelect HSS C18 SB VanGuard Cartridge, 100Å, 3.5 µm, 2.1 mm X 5 mm 1 * 3 items
Catalog # WATE186007842
Supplier:  WATERS
XSelect HSS C18 SB VanGuard Cartridge, 100Å, 3.5 µm, 2.1 mm X 5 mm 1 * 3 items
Catalog # WATE186007842
Supplier:  WATERS
Frequently Bought Together